Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172107.1 | NP_001165578.1 | 4 | 481 | ||
NM_153528.2 | NP_705756.1 | 4 | 711 | |||
GenBank mRNA | AK029543.1 | - | 4 | 482 | ||
AK035257.1 | - | 4 | 714 | |||
AK129016.1 | - | 4 | 650 | |||
BC037472.1 | - | 4 | 645 | 4236901 |