Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172157.1 | NP_001165628.1 | 14 | 1663 | ||
NM_153530.2 | NP_705758.1 | 13 | 1621 | |||
GenBank mRNA | AK029652.1 | - | 14 | 1677 | ||
AK031180.1 | - | 14 | 1646 | |||
AK183644.1 | - | 1 | 17 | |||
BC036177.1 | - | 13 | 1582 | 5004341 |