Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sned1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172463.4 | NP_766051.4 | 1, 2 | 305 | ||
GenBank mRNA | AJ584850.1 | - | 1, 2 | 361 | ||
AK086298.1 | - | 1, 2 | 362 | |||
AY169783.1 | - | 1, 2 | 360 | |||
BC145886.1 | - | 1, 2 | 305 | 40131189 |