Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000284.3 | NP_000275.1 | 6 | 685 | ||
NM_001173454.1 | NP_001166925.1 | 7 | 799 | |||
NM_001173455.1 | NP_001166926.1 | 6 | 706 | |||
GenBank mRNA | AK129887.1 | - | 6 | 620 | ||
AK222740.1 | - | 6 | 616 | |||
AK222988.1 | - | 6 | 650 | |||
AK293250.1 | - | 7 | 764 | |||
AK296341.1 | - | 6 | 672 | |||
AK312263.1 | - | 6 | 651 | |||
BC002406.1 | - | 6 | 597 | 2961286 | ||
EF576990.1 | - | 3 | 154 | |||
EF590117.1 | - | 717 | ||||
J03503.1 | - | 6 | 612 | |||
J03575.1 | - | 6 | 585 | |||
L13318.1 | - | 6 | 645 | |||
L48690.1 | - | 6 | 645 | |||
M24848.1 | - | 6 | 583 | |||
X52709.1 | - | 6 | 594 | |||
X52710.1 | - | 6 | 596 |