Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zswim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172503.3 | NP_766091.2 | 3 | 397 | ||
GenBank mRNA | AK041934.1 | - | 3 | 411 | ||
AK052168.1 | - | 3 | 411 | |||
AK053663.1 | - | 3 | 411 | |||
AK150221.1 | - | 3 | 411 | |||
BC058080.1 | - | 3 | 397 | 6822376 |