Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285928.1 | NP_001272857.1 | 4 | 815 | ||
NM_001285929.1 | NP_001272858.1 | 3 | 704 | |||
NM_001285930.1 | NP_001272859.1 | 3 | 568 | |||
NM_178671.5 | NP_848786.1 | 2 | 437 | |||
GenBank mRNA | AK045964.1 | - | 2 | 395 | ||
AK046949.1 | - | 4 | 816 | |||
AK077656.1 | - | 2 | 439 | |||
AK080662.1 | - | 2 | 403 | |||
AK133185.1 | - | 2 | 423 | |||
AK161245.1 | - | 2 | 424 | |||
AK161296.1 | - | 2 | 400 |