Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033531.3 | NP_001028703.2 | 6 | 1215 | ||
NM_001163020.1 | NP_001156492.1 | 7 | 1314 | |||
GenBank mRNA | AK122569.1 | - | 931 | |||
AK134944.1 | - | 6 | 1187 | |||
AK139083.1 | - | 2 | 556 | |||
BC141239.1 | - | 6 | 919 | 9055851 | ||
BC145566.1 | - | 5 | 811 |