Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146184.1 | NP_001139656.1 | 9 | 3850 | ||
NM_001146185.1 | NP_001139657.1 | 9 | 3536 | |||
NM_001146186.1 | NP_001139658.1 | 7 | 3883 | |||
NM_001146187.1 | NP_001139659.1 | 9 | 3542 | |||
NM_006210.2 | NP_006201.1 | 10 | 3926 | |||
GenBank mRNA | AB003039.1 | - | 9 | 3499 | ||
AB006625.2 | - | 9 | 3515 | |||
AF208967.1 | - | 10 | 3896 | |||
AF208968.1 | - | 9 | 3820 | |||
AF208969.1 | - | 9 | 3506 | |||
AF208970.1 | - | 7 | 3884 | |||
AK295679.1 | - | 9 | 3542 | |||
BC136268.1 | - | 8 | 3370 | 9020255 | ||
BC150272.1 | - | 9 | 3515 | 9007244 | ||
CR933682.1 | - | 1 | 1193 | |||
U90336.1 | - | 1 | 613 |