Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081230.1 | NP_001074699.1 | 7 | 1127 | ||
GenBank mRNA | AK030527.1 | - | 7 | 968 | ||
AK038737.1 | - | 7 | 1072 | |||
AK135702.1 | - | 4 | 621 | |||
AK135823.1 | - | 7 | 1031 | |||
AK138540.1 | - | 7 | 1072 | |||
AK147287.1 | - | 7 | 1070 | |||
AK147416.1 | - | 7 | 1070 | |||
AK162555.1 | - | 7 | 1047 | |||
BC061216.1 | - | 7 | 983 | |||
BC087935.1 | - | 7 | 1024 |