Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrtm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178678.4 | NP_848793.1 | 2 | 1158 | ||
GenBank mRNA | AK032485.1 | - | 2 | 1175 | ||
AK035773.1 | - | 2 | 1155 | |||
AK035858.1 | - | 2 | 1161 | |||
AK035882.1 | - | 2 | 1159 | |||
AK036180.1 | - | 2 | 1178 | |||
AK036366.1 | - | 2 | 1152 | |||
AK163753.1 | - | 2 | 1151 | |||
AY182029.1 | - | 1 | 616 | |||
BC094910.1 | - | 2 | 645 | 30044805 | ||
BC113178.1 | - | 2 | 832 | 40060681 | ||
BC113784.1 | - | 2 | 832 | 40060682 |