Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172563.3 | NP_766151.1 | 3 | 1191 | ||
GenBank mRNA | AK054062.1 | - | 3 | 681 | ||
AK079068.1 | - | 4 | 652 | |||
AK079308.1 | - | 3 | 566 | |||
AK179453.1 | - | 1 | 103 | |||
BC057693.1 | - | 2 | 609 | 4196259 | ||
BC058705.1 | - | 3 | 1169 | 6839029 |