Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304451.1 | NP_001291380.1 | 9 | 1081 | ||
NM_001304452.1 | NP_001291381.1 | 10 | 1226 | |||
NM_002631.3 | NP_002622.2 | 10 | 1147 | |||
GenBank mRNA | AK290404.1 | - | 10 | 1072 | ||
AK296889.1 | - | 11 | 975 | |||
AK298830.1 | - | 10 | 1226 | |||
AK300952.1 | - | 9 | 984 | |||
AK303898.1 | - | 8 | 734 | |||
AK304423.1 | - | 9 | 1004 | |||
BC000368.2 | - | 10 | 1058 | 2819330 | ||
U30255.1 | - | 9 | 1027 |