Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145471.2 | NP_663446.1 | 4 | 638 | ||
GenBank mRNA | AK036278.1 | - | 2 | 511 | ||
AK047414.1 | - | 2 | 598 | |||
AK053747.1 | - | 568 | ||||
AK132158.1 | - | 2 | 1202 | |||
AK149655.1 | - | 2 | 600 | |||
AK159836.1 | - | 2 | 581 | |||
AK172871.1 | - | 2 | 1181 | |||
AK191794.1 | - | 1 | 130 | |||
AY239225.1 | - | 3 | 689 | |||
AY239226.1 | - | 4 | 551 | |||
AY344668.1 | - | 2 | 600 | |||
BC022126.1 | - | 4 | 671 | 5097371 | ||
BC033482.1 | - | 2 | 425 | |||
BC138217.1 | - | 2 | 812 | 8861236 | ||
BC145203.1 | - | 2 | 812 |