Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C030006K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145472.2 | NP_663447.1 | 1 | 163 | ||
NM_176828.4 | NP_789798.2 | 1 | 163 | |||
GenBank mRNA | AK021050.1 | - | 1 | 152 | ||
AK046662.1 | - | 1 | 131 | |||
AK053546.1 | - | 1 | 141 | |||
AK075645.1 | - | 1 | 159 | |||
AK085641.1 | - | 1 | 159 | |||
AK140368.1 | - | 1 | 159 | |||
AK140578.1 | - | 1 | 164 | |||
AK163615.1 | - | 1 | 146 | |||
AK165937.1 | - | 1 | 157 | |||
BC019462.1 | - | 1 | 139 | 4458439 | ||
BC026997.1 | - | 1 | 137 | 5362226 |