Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182928.4 | NP_891558.1 | 3 | 830 | ||
GenBank mRNA | AB121035.1 | - | 3 | 754 | ||
BB705319.1 | - | 1 | 207 | |||
BC096428.1 | - | 3 | 757 | 4948766 | ||
DV649647.1 | - | 3 | 706 |