Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178923.3 | NP_849254.2 | 3 | 524 | ||
GenBank mRNA | AK045266.1 | - | 3 | 521 | ||
AK048239.1 | - | 3 | 586 | |||
AK173120.1 | - | 3 | 602 | |||
BC053096.1 | - | 3 | 419 | 6401143 | ||
BC057592.1 | - | 3 | 524 | 6417362 |