Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122752.1 | NP_001116224.1 | 7, 8 | 1272 | ||
NM_005025.4 | NP_005016.1 | 7, 8 | 1505 | |||
XM_017006618.1 | XP_016862107.1 | 7, 8 | 1570 | |||
GenBank mRNA | AK290082.1 | - | 7, 8 | 1195 | ||
BC018043.1 | - | 7, 8 | 1193 | 4796470 | ||
CR627434.1 | - | 7, 8 | 1494 | |||
KU178259.1 | - | 6, 7 | 1074 | |||
KU178260.1 | - | 3, 4 | 455 | |||
Z81326.1 | - | 7, 8 | 1155 |