Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028639.3 | NP_082915.1 | 1, 2 | 531 | ||
GenBank mRNA | AK040578.1 | - | 1, 2 | 534 | ||
AK040610.1 | - | 1, 2 | 501 | |||
AK050263.1 | - | 1, 2 | 518 | |||
AK080284.1 | - | 1, 2 | 510 | |||
AK153704.1 | - | 1, 2 | 513 | |||
BC042512.1 | - | 1, 2 | 208 | 4223801 |