Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145514.5 | NP_663489.4 | 10 | 2037 | ||
GenBank mRNA | AK075743.1 | - | 10 | 1429 | ||
AK082326.1 | - | 11 | 1332 | |||
AK213047.1 | - | 1 | 29 | |||
AK218758.1 | - | 1 | 29 | |||
BC020044.1 | - | 6 | 782 | |||
BC058601.1 | - | 8 | 1033 | |||
BC094233.1 | - | 7 | 921 | |||
BC139336.1 | - | 10 | 1199 | 8862358 | ||
BC139337.1 | - | 10 | 1199 | 8862359 |