Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172684.2 | NP_766272.2 | 2 | 1409 | ||
GenBank mRNA | AB101658.1 | - | 2 | 1411 | ||
AK087685.1 | - | 1 | 375 | |||
AK134093.1 | - | 2 | 410 | |||
AK138603.1 | - | 2 | 953 | |||
BC094667.1 | - | 2 | 934 |