Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080771.1 | NP_001074240.1 | 2 | 563 | ||
GenBank mRNA | AK149252.1 | - | 2 | 446 | ||
BB665287.1 | - | 2 | 383 | |||
BC017632.1 | - | 2 | 563 |