Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033306.1 | NP_001028478.1 | 3, 4 | 1615 | ||
GenBank mRNA | AK145414.1 | - | 3, 4 | 474 | ||
BC028832.1 | - | 2, 3 | 477 | |||
BC048850.1 | - | 2, 3 | 482 | |||
BC058986.1 | - | 3, 4 | 1117 |