Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081237.1 | NP_001074706.1 | 1, 2 | 1296 | ||
GenBank mRNA | AK048908.1 | - | 1, 2 | 885 | ||
AK052765.1 | - | 1, 2 | 873 | |||
AK083047.1 | - | 1, 2 | 881 | |||
AK129335.1 | - | 1, 2 | 206 | |||
BC037084.1 | - | 1, 2 | 901 | |||
BC147633.1 | - | 1, 2 | 1052 | 9056623 | ||
BC147636.1 | - | 1, 2 | 1051 | 9056626 | ||
BC151064.1 | - | 1, 2 | 1052 | 9087977 | ||
BC151065.1 | - | 1, 2 | 1052 | 9087978 | ||
BC151068.1 | - | 1, 2 | 1052 | 9087981 | ||
BC151076.1 | - | 1, 2 | 1052 | 9087989 |