Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blvrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290525.1 | NP_001277454.1 | 3 | 310 | ||
NM_144923.3 | NP_659172.1 | 3 | 396 | |||
GenBank mRNA | AK153153.1 | - | 3 | 317 | ||
BC006617.1 | - | 3 | 336 | 3967600 | ||
BC027279.1 | - | 3 | 396 | 3492861 |