Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000672.3 | NP_000663.1 | 8 | 1145 | ||
NM_001102470.1 | NP_001095940.1 | 8 | 1145 | |||
NR_132990.1 | 6 | 786 | ||||
GenBank mRNA | AJ278908.1 | - | 1 | 79 | ||
AK092768.1 | - | 8 | 1145 | |||
AK298290.1 | - | 4 | 714 | |||
AK316326.1 | - | 6 | 714 | |||
BC039065.1 | - | 7 | 998 | 5184104 |