Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033877.4 | NP_001029049.2 | 10 | 1440 | ||
GenBank mRNA | AK053131.1 | - | 10 | 1441 | ||
BC141170.1 | - | 10 | 1392 | |||
CF741491.1 | - | 2 | 182 | |||
CX769149.1 | - | 2 | 143 |