Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hykk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177351.4 | NP_796325.2 | 2 | 758 | ||
GenBank mRNA | AK048771.1 | - | 2 | 731 | ||
AK083218.1 | - | 1 | 3352 | |||
AK085527.1 | - | 2 | 767 | |||
AK138851.1 | - | 2 | 744 | |||
AK142509.1 | - | 1 | 3354 | |||
AK162425.1 | - | 2 | 759 | |||
AK163926.1 | - | 2 | 738 | |||
BC038618.1 | - | 2 | 737 | 4221933 |