Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kbtbd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008785.5 | NP_001008785.2 | 3 | 592 | ||
NM_001102670.2 | NP_001096140.1 | 2 | 381 | |||
GenBank mRNA | AK142080.1 | - | 3 | 566 | ||
AK167507.1 | - | 2 | 332 | |||
BC088734.1 | - | 2 | 503 | 30459462 |