Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172904.2 | NP_766492.2 | 2 | 566 | ||
GenBank mRNA | AK028142.1 | - | 2 | 558 | ||
AK036674.1 | - | 532 |