Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162492.1 | NP_001155964.1 | 2 | 382 | ||
NM_001162493.1 | NP_001155965.1 | 2 | 352 | |||
NM_138670.3 | NP_619611.3 | 2 | 486 | |||
GenBank mRNA | AK136571.1 | - | 1 | 381 | ||
BC004079.1 | - | 1 | 327 | 3592659 | ||
BC006066.1 | - | 1 | 327 | |||
BC094469.1 | - | 2 | 486 | 5009481 |