Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145211.2 | NP_660212.2 | 4 | 755 | ||
GenBank mRNA | AK149613.1 | - | 4 | 716 | ||
BC013715.1 | - | 4 | 737 | 3257027 | ||
BC057878.1 | - | 4 | 714 | 5348858 | ||
M33863.1 | - | 741 | ||||
X04958.1 | - | 4 | 741 |