Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011736.1 | NP_001011736.1 | 1 | 476 | ||
GenBank mRNA | BC150839.1 | - | 1 | 501 | 9087750 |
Gene Symbol: Olfr204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146992.2 | NP_667203.2 | 1 | 476 |