Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Robo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175549.4 | NP_780758.3 | 2 | 358 | ||
GenBank mRNA | AK031495.1 | - | 2 | 884 | ||
AK035110.1 | - | 2 | 1232 | |||
AK048081.1 | - | 2 | 1272 | |||
AK051227.1 | - | 2 | 1272 | |||
AK085461.1 | - | 2 | 983 | |||
AK164874.1 | - | 2 | 982 | |||
BC055333.1 | - | 2 | 666 | 6409283 | ||
DQ533875.1 | - | 3 | 638 | |||
DQ533876.1 | - | 2 | 358 |