Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mon1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048143.2 | NP_001041608.1 | 4 | 714 | ||
NM_173015.3 | NP_766603.1 | 3 | 904 | |||
GenBank mRNA | AK030288.1 | - | 3 | 870 | ||
AK157390.1 | - | 4 | 714 | |||
AK173047.1 | - | 3 | 576 | |||
BC062639.1 | - | 3 | 580 | |||
CA320533.1 | - | 3 | 494 | |||
CJ089261.1 | - | 4 | 715 |