Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002239.3 | NP_001002239.2 | 7 | 678 | ||
GenBank mRNA | AK011132.1 | - | 7 | 634 | ||
AK011133.1 | - | 7 | 634 |