Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138590.4 | NP_613056.2 | 4 | 841 | ||
GenBank mRNA | AK029808.1 | - | 4 | 848 | ||
AK032353.1 | - | 4 | 813 | |||
AK039158.1 | - | 4 | 762 | |||
AK043238.1 | - | 4 | 754 | |||
AK080785.1 | - | 5 | 930 | |||
AK157728.1 | - | 4 | 744 | |||
BC050200.1 | - | 3 | 716 | |||
BC117020.1 | - | 4 | 744 | 40126339 | ||
BC117022.1 | - | 4 | 744 | 40126341 | ||
BC144774.1 | - | 4 | 748 | 9053322 | ||
CF732773.1 | - | 3 | 432 |