Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177214.4 | NP_796188.2 | 42 | 6134 | ||
GenBank mRNA | AK149291.1 | - | 13 | 1657 | ||
AK173025.1 | - | 37 | 5363 | |||
BC011390.1 | - | 15 | 2323 | |||
BC023685.1 | - | 15 | 2248 | |||
BC030856.1 | - | 14 | 2075 | |||
BC055940.1 | - | 6 | 922 | |||
BC058608.1 | - | 11 | 1617 | |||
BC063261.1 | - | 42 | 6134 | 6408607 |