Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145978.2 | NP_001139450.2 | 25 | 3122 | ||
GenBank mRNA | AK054229.1 | - | 25 | 3086 | ||
AK164513.1 | - | 25 | 3134 | |||
AK172907.1 | - | 25 | 3054 | |||
BC046397.1 | - | 11 | 1143 |