Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsper4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130030.1 | NP_001123502.1 | 8 | 999 | ||
NM_177866.4 | NP_808534.1 | 10 | 1218 | |||
GenBank mRNA | AK077145.1 | - | 10 | 1205 | ||
BC108974.2 | - | 10 | 1198 | 40047424 | ||
BC108975.1 | - | 10 | 1198 | 40047425 | ||
EU781510.1 | - | 9 | 1095 | |||
EU781511.1 | - | 8 | 974 |