Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033425.4 | NP_001028597.2 | 6 | 1809 | ||
NM_001289481.1 | NP_001276410.1 | 7 | 1713 | |||
NM_001289482.1 | NP_001276411.1 | 4 | 1479 | |||
NM_001289483.1 | NP_001276412.1 | 7 | 1209 | |||
NM_001289484.1 | NP_001276413.1 | 7 | 1578 | |||
GenBank mRNA | AK141259.1 | - | 6 | 1780 | ||
BC145638.1 | - | 6 | 1546 | 9054214 | ||
BI133651.1 | - | 1 | 400 | |||
DQ323929.1 | - | 6 | 1777 | |||
DQ323930.1 | - | 7 | 1681 | |||
DQ323931.1 | - | 4 | 1447 | |||
EF152498.1 | - | 5 | 1624 |