Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207261.3 | NP_997144.1 | 3 | 816 | ||
GenBank mRNA | AK139600.1 | - | 3 | 1250 | ||
AK162136.1 | - | 3 | 819 | |||
AY325301.1 | - | 3 | 730 | |||
AY542902.1 | - | 3 | 730 | |||
BC104132.1 | - | 3 | 730 | |||
BC104133.1 | - | 3 | 727 | |||
BC115705.1 | - | 3 | 729 | |||
BC115887.1 | - | 3 | 723 | |||
BC127136.1 | - | 3 | 730 | 40045157 | ||
BC127137.1 | - | 3 | 730 | 40045158 |