Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dppa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163358.1 | NP_001156830.1 | 3 | 151 | ||
NM_178247.3 | NP_839978.1 | 4 | 237 | |||
GenBank mRNA | AF490345.1 | - | 4 | 223 | ||
AK145642.1 | - | 4 | 238 | |||
AK166752.1 | - | 4 | 235 | |||
BC115806.1 | - | 3 | 178 | 40106955 | ||
BC115901.1 | - | 3 | 175 | 40106956 |