Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183034.1 | NP_898855.1 | 7 | 2066 | ||
GenBank mRNA | AK154703.1 | - | 6 | 1995 | ||
AK155357.1 | - | 7 | 2090 | |||
AK157752.1 | - | 6 | 1710 | |||
BC053079.1 | - | 7 | 2066 | 5702529 |