Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A230065H16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101503.1 | NP_001094973.1 | 4 | 476 | ||
GenBank mRNA | AK038816.1 | - | 4 | 476 |