Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007463.1 | NP_001007464.1 | 5, 6 | 1167 | ||
NM_001290462.1 | NP_001277391.1 | 5, 6 | 1167 | |||
GenBank mRNA | AK132968.1 | - | 5, 6 | 1168 | ||
AY769082.1 | - | 5, 6 | 1167 | |||
BC150868.1 | - | 5, 6 | 1209 | 9087779 |