Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aym1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012726.2 | NP_001012744.2 | 1 | 442 | ||
GenBank mRNA | AF069744.1 | - | 1 | 448 |