Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009456.2 | NP_033482.1 | 2 | 92 | ||
GenBank mRNA | AJ130961.1 | - | 2 | 81 | ||
AK152522.1 | - | 2 | 94 | |||
AK165620.1 | - | 2 | 73 | |||
AK167847.1 | - | 2 | 80 | |||
BC049075.1 | - | 2 | 79 | |||
BC057959.1 | - | 2 | 73 | |||
BC093503.1 | - | 2 | 71 | 4216580 | ||
BC106149.1 | - | 2 | 70 | 4502101 | ||
X97042.1 | - | 2 | 92 |