Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ROBO2 SIRNA Assays ›
Gene Symbol:
ROBO2
Gene Name:
roundabout guidance receptor 2
Gene Aliases:
SAX3
Chromosome Location:
Chr.3: 75906675 - 77649964 on Build GRCh38
UniGene ID:
Hs.13305
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s12095
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ROBO2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128929.3
NP_001122401.1
26
4429
NM_001290039.1
NP_001276968.1
26
4737
NM_001290040.1
NP_001276969.1
27
4920
NM_001290065.1
NP_001276994.1
26
4864
NM_002942.4
NP_002933.1
25
4725
XM_011533981.2
XP_011532283.1
29
5144
XM_011533982.1
XP_011532284.1
28
5018
XM_011533983.1
XP_011532285.1
28
4961
XM_011533984.1
XP_011532286.1
27
4688
XM_011533985.1
XP_011532287.1
26
4562
XM_017006986.1
XP_016862475.1
29
5367
XM_017006987.1
XP_016862476.1
28
5009
XM_017006988.1
XP_016862477.1
28
4961
XM_017006989.1
XP_016862478.1
27
4949
XM_017006990.1
XP_016862479.1
29
4840
XM_017006991.1
XP_016862480.1
27
4835
XM_017006992.1
XP_016862481.1
28
4714
XM_017006993.1
XP_016862482.1
26
5046
XM_017006994.1
XP_016862483.1
27
4688
XM_017006995.1
XP_016862484.1
27
4679
XM_017006996.1
XP_016862485.1
28
4567
XM_017006997.1
XP_016862486.1
28
4567
XM_017006998.1
XP_016862487.1
27
4911
XM_017006999.1
XP_016862488.1
27
4911
XM_017007000.1
XP_016862489.1
26
4899
XM_017007001.1
XP_016862490.1
26
4562
XM_017007002.1
XP_016862491.1
26
4553
XM_017007003.1
XP_016862492.1
25
4550
XM_017007004.1
XP_016862493.1
27
4441
XM_017007005.1
XP_016862494.1
27
4441
XM_017007006.1
XP_016862495.1
26
4776
GenBank mRNA
AB046788.1
-
25
4088
BC144303.1
-
27
4333
BC146772.1
-
25
4088
9007219
BX648828.1
-
2
6070
DQ533873.1
-
26
4410
DQ533874.1
-
25
4725
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
metanephros development
ureteric bud development
homophilic cell adhesion via plasma membrane adhesion molecules
axon guidance
central nervous system development
brain development
axon midline choice point recognition
olfactory bulb interneuron development
retinal ganglion cell axon guidance
cellular response to hormone stimulus
Roundabout signaling pathway
positive regulation of axonogenesis
negative regulation of negative chemotaxis
negative regulation of synapse assembly
apoptotic process involved in luteolysis
protein binding
axon guidance receptor activity
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0