Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lekr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037923.4 | NP_001033012.2 | 3 | 353 | ||
NM_001166659.1 | NP_001160131.1 | 3 | 353 | |||
GenBank mRNA | AK137452.1 | - | 3 | 313 | ||
BC147278.1 | - | 2 | 282 | 9056357 | ||
BC147279.1 | - | 2 | 282 | 9056330 | ||
CJ055787.1 | - | 3 | 348 | |||
CJ137734.1 | - | 3 | 354 |