Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twistnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172253.2 | NP_758457.1 | 4 | 820 | ||
GenBank mRNA | AK008677.1 | - | 4 | 802 | ||
AK010829.1 | - | 4 | 821 | |||
AK016638.1 | - | 4 | 817 | |||
AK017016.1 | - | 4 | 808 | |||
AK153090.1 | - | 810 | ||||
BC082580.1 | - | 792 | ||||
BC139162.1 | - | 4 | 808 | 8862184 | ||
BC139163.1 | - | 4 | 808 | 8862185 |